Learn More
Apexbio Technology LLC Fosamprenavir Calcium Salt 226700-81-8 5mg
SDP
Supplier:  Apexbio Technology LLC A34215
Encompass_Preferred
Fosamprenavir Calcium Salt (CAS 226700-81-8) is a small-molecule inhibitor targeting HIV-1 protease It is designed to selectively inhibit HIV-1 protease thereby preventing cleavage of viral polyprotein precursors required for mature virion assembly and viral replication Fosamprenavir Calcium Salt exerts its biological activity primarily through competitive inhibition of HIV-1 protease In research studies Fosamprenavir Calcium Salt demonstrates inhibitory activity with reported IC50 values against HIV-1 protease in the nanomolar range Based on these pharmacological properties Fosamprenavir Calcium Salt holds research potential in studies on antiretroviral combination therapies bioavailability safety profiles and pharmacodynamic assessments targeting viral resistance mechanisms
Catalog No. 50-000-92878
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.