Learn More
Apexbio Technology LLC Calcifediol monohydrate 63283-36-3 5mg
SDP
Supplier:  Apexbio Technology LLC A32705
Encompass_Preferred
Calcifediol monohydrate (63283-36-3) is a precursor to the active vitamin D hormone that acts via the vitamin D receptor (VDR) It is designed to modulate vitamin D signaling thereby regulating pathways involved in calcium homeostasis and cellular growth Calcifediol monohydrate exerts its biological activity primarily through VDR activation In bovine parathyroid cells calcifediol reduces parathyroid hormone (PTH) secretion and mRNA expression Studies in T-47D breast cancer cells demonstrate that calcifediol s function involves VDR activation linked to endocytic uptake of vitamin D-binding protein (DBP) Based on these pharmacological properties calcifediol monohydrate holds research potential in studies assessing vitamin D s role in diseases such as colorectal breast ovarian renal pancreatic and prostate cancers
Catalog No. 50-000-92879
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.