Learn More
Apexbio Technology LLC Fosaprepitant dimeglumine salt 265121-04-8 50mg
SDP
Supplier:  Apexbio Technology LLC B214550
Encompass_Preferred
Fosaprepitant dimeglumine salt (CAS 265121-04-8) also known as MK-0517 or L-758 298 is a selective antagonist of the neurokinin-1 (NK-1) receptor It functions primarily through its active metabolite aprepitant a brain-penetrant NK-1 inhibitor with high affinity thereby effectively suppressing substance P-mediated signaling In preclinical models fosaprepitant dimeglumine salt demonstrates antiemetic activity against cisplatin-induced vomiting In biomedical research it is often investigated alongside corticosteroids (e g dexamethasone) and 5-HT3 antagonists (e g granisetron) for the management of chemotherapy-induced nausea and vomiting (CINV)
Catalog No. 50-000-63700
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.