Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Fosetyl-aluminum

Catalog No. AC467281000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467281000 100 g
AC467280250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467281000 Supplier Thermo Scientific Chemicals Supplier No. 467281000
  • $329.00
    $238.65 / Each of 1

    Save $90.35

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 39148-24-8 | C6H15AlO9P3, C6H18AlO9P3 | 351.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39148-24-8
Molecular Formula C6H18AlO9P3

Specifications

Quantity 100 g
Solubility Solubility aq. soln.: 120 g/L (20°C). Other solubilities: practically insoluble in acetonitrile, propylene, glycol
Melting Point >300.0°C
Formula Weight 354.11
Chemical Name or Material Fosetyl-Aluminum
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes serious eye damage.

Precautionary Statement GHS P Statement Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 254-320-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.