Learn More
Description
- For antimicrobial susceptibility testing of mecillinam as recommended by CLSI and EUCAST standards
Specifications
Specifications
| Format | Paper Disc |
| Product Type | Antimicrobial Susceptibility Disc |
| Target Organism Class | Gram-negative Microorganisms, Gram-positive Microorganisms, Enterobacterales, Enterococcus |
| Test Type | Antimicrobial Susceptibility |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
