Learn More
Foss™ Dummy Tubes
Supplier:  Foss™ 500059
 View more versions of this product
Catalog No. TC500059
Description
Description
Specifications
Specifications
Dummy Tube
Exhaust manifold when less than 8 or 20 samples are being digested
Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.