Learn More
Foss™ Cellulose Thimbles
Supplier: Foss™ 15220053
This item is not returnable.
View return policy
Description
Cellulose Thimbles, 28 × 22mm. For use with 2047 SoxCap and Soxtec 2045, 2050, and 2055. 25 per package.
Specifications
Cellulose | |
For fat analysis | |
For use with 2047 SoxCap and Soxtec 2045, 2050, and 2055 | |
Extraction Thimble |