Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Fossil Replica Collection, 4 Pieces
SCI_ED

Catalog No. S15015 Shop All Eisco Products
Change view
Click to view available options
Grade:
K to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S15015 K to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15015 Supplier Eisco™ Supplier No. BGFOSSET4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Fossil Replica Collection includes four identified specimens, information cards, and a geological timescale.

  • Set of 4 replica fossils - Bivalve, Fish, Hardouinia, Perisphinctes
  • Each fossil comes packaged with an information card in a plastic bag for safe and convenient storage
  • Use the included timescale to locate fossil specimens in geologic history
  • Ideal in an earth science classroom, or for any geologist's collection
  • Assembled in the United States

Specifications

Grade K to 14
Age 5 to 20
For Use With (Application) Fossil Identification
Includes Bivalve, Fish, Hardouinia, Perisphinctes, Information Card, and Timeline
Product Type Fossils
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.