Learn More
Fotodyne™ FOTO/Analyst™ Investigator Eclipse UV Workstations: 0.4 MPix USB Camera
Catalog No. FD61510UVNP
Change view
Click to view available options
Printer Included:
No
Yes
Transilluminator:
Dual wavelength UV
Single wavelength UV
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Printer Included Transilluminator
FD61510UVNP No Single wavelength UV
02-580-150 No Dual wavelength UV
02-580-149 Yes Dual wavelength UV
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FD61510UVNP Supplier Fotodyne™ Supplier No. 61510UVNP
Easy-to-use gel doc system quickly captures digital images of gels and saves them as TIF, BMP and JPG formats
Offer space-saving FOTO/Eclipse benchtop darkroom plus gliding transilluminator platform for easy access to gels, epi-white light LEDs for framing, focusing and opaque samples, and safety-interlocking door to prevent accidental UV exposure.
  • Live-image feature to adjust camera settings while viewing real-time changes on a full-size computer monitor
  • Flexible integration feature offers continuously adjustable exposure settings rather than a limited number of presets
  • Control to optimize image quality while maximizing sensitivity
  • As little as 0.1ng of ethidium bromide-stained DNA can be detected
  • Show saturation, crop, label lanes and bands, draw boxes and arrows, and more
  • Scientific grade CCD camera with zoom lens and exposure control in 0.1 second increments
  • Safety interlock door can be manually overridden to allow band cutting
  • Constructed of sturdy 16-gauge steel
Specifications
For Use With (Equipment) Basic 1D Gels
Transilluminator Single wavelength UV
Printer Included No
For Use With (Application) Basic 1D Gels
Product Type UV Workstation