Learn More
Fotodyne™ FOTO/Analyst™ Investigator Eclipse UV Workstations: 2.0MPix Camera
Catalog No. FD61710UV
Change view
Click to view available options
For Use With (Application):
1D Complex Gels
2D Complex Gels
Printer Included:
No
Yes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Printer Included
FD61710UV 1D Complex Gels Yes
FD61710S2UV 2D Complex Gels Yes
61-710-S2UVNP 2D Complex Gels No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FD61710UV Supplier Fotodyne™ Supplier No. 61710UV
Easy-to-use gel doc system quickly captures digital images of gels and saves them as TIF, BMP and JPG formats
Offer space-saving FOTO/Eclipse benchtop darkroom plus gliding transilluminator platform for easy access to gels, epi-white light LEDs for framing, focusing and opaque samples, and safety-interlocking door to prevent accidental UV exposure.
  • Live-image feature to adjust camera settings while viewing real-time changes on a full-size computer monitor
  • Flexible integration feature offers continuously adjustable exposure settings rather than a limited number of presets
  • Control to optimize image quality while maximizing sensitivity
  • As little as 0.1ng of ethidium bromide-stained DNA can be detected
  • Show saturation, crop, label lanes and bands, draw boxes and arrows, and more
  • Scientific grade CCD camera with zoom lens and exposure control in 0.1 second increments
  • Safety interlock door can be manually overridden to allow band cutting
  • Constructed of sturdy 16-gauge steel
Specifications
For Use With (Equipment) 1D Complex Gels
Transilluminator Dual Wavelength UV
Printer Included Yes
Camera Type 2MP USB
For Use With (Application) 1D Complex Gels
Product Type UV Workstation