Learn More
Fotodyne™ FOTO/Analyst™ Investigator Eclipse UV Workstations: 1.4 MPix Camera
Description
Offer space-saving FOTO/Eclipse benchtop darkroom plus gliding transilluminator platform for easy access to gels, epi-white light LEDs for framing, focusing and opaque samples, and safety-interlocking door to prevent accidental UV exposure.
- Live-image feature to adjust camera settings while viewing real-time changes on a full-size computer monitor
- Flexible integration feature offers continuously adjustable exposure settings rather than a limited number of presets
- Control to optimize image quality while maximizing sensitivity
- As little as 0.1ng of ethidium bromide-stained DNA can be detected
- Show saturation, crop, label lanes and bands, draw boxes and arrows, and more
- Scientific grade CCD camera with zoom lens and exposure control in 0.1 second increments
- Safety interlock door can be manually overridden to allow band cutting
- Constructed of sturdy 16-gauge steel
Specifications
Specifications
| For Use With (Equipment) | 1D or 2D Complex Gels |
| Transilluminator | Single wavelength UV |
| Printer Included | Yes |
| For Use With (Application) | 1D or 2D Complex Gels |
| Product Type | UV Workstation |