Learn More
Fotodyne™ FOTO/Analyst™ Investigator Eclipse UV Workstations: 1.4 MPix Camera
Catalog No. FD61610UVNP
Change view
Click to view available options
Printer Included:
No
Yes
Transilluminator:
Dual wavelength UV
Single wavelength UV
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Printer Included Transilluminator
FD61610UVNP No Single wavelength UV
FD61610UV Yes Single wavelength UV
02-580-153 Yes Dual wavelength UV
02-580-154 No Dual wavelength UV
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FD61610UVNP Supplier Fotodyne™ Supplier No. 61610UVNP
Easy-to-use gel doc system quickly captures digital images of gels and saves them as TIF, BMP and JPG formats
Offer space-saving FOTO/Eclipse benchtop darkroom plus gliding transilluminator platform for easy access to gels, epi-white light LEDs for framing, focusing and opaque samples, and safety-interlocking door to prevent accidental UV exposure.
  • Live-image feature to adjust camera settings while viewing real-time changes on a full-size computer monitor
  • Flexible integration feature offers continuously adjustable exposure settings rather than a limited number of presets
  • Control to optimize image quality while maximizing sensitivity
  • As little as 0.1ng of ethidium bromide-stained DNA can be detected
  • Show saturation, crop, label lanes and bands, draw boxes and arrows, and more
  • Scientific grade CCD camera with zoom lens and exposure control in 0.1 second increments
  • Safety interlock door can be manually overridden to allow band cutting
  • Constructed of sturdy 16-gauge steel
Specifications
For Use With (Equipment) 1D or 2D Complex Gels
Transilluminator Single wavelength UV
Printer Included No
For Use With (Application) 1D or 2D Complex Gels
Product Type UV Workstation