Learn More
Fotodyne™ FOTO/Analyst™ Investigator/FX Workstations with Manual Zoom, Side Lights, Single Wavelength
Catalog No. 71670UVNP
Change view
Click to view available options
Single-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 1.4 Mpx camera
Single-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 2.0 Mpx camera
Single-wavelength UV transilluminator; w/PC; 8-bit, 1.4 Mpx camera
Single-wavelength UV transilluminator; w/PC; 8-bit, 2.0 Mpx camera
Single-wavelength UV, dual-light transilluminator; w/PC and thermal printer; 12-bit, 1.4 Mpx camera
Single-wavelength UV, dual-light transilluminator; w/PC, thermal printer; 8-bit, 1.4 Mpx cam
Single-wavelength UV, dual-light transilluminator; w/PC, thermal printer; 8-bit, 2.0 Mpx
Single-wavelength UV, dual-light transilluminator; w/PC, w/o printer; 12-bit, 1.4 Mpx camera
Single-wavelength UV, dual-light transilluminator; w/PC, w/o printer; 8-bit, 2.0 Mpx camera
Single-wavelength UV, dual-light transilluminator; w/PC; 8-bit, 1.4 Mpx camera
Single-wavelength UV, dual-light transilluminator; w/PC; 8-bit, 2.0 Mpx camera
UV transilluminator; w/PC and thermal printer; 8-bit, 0.4 Mpx camera
UV transilluminator; w/PC, w/o printer; 8-bit, 0.4 Mpx camera
UV transilluminator; w/PC, w/o printer; 8-bit, 2.0Mpx camera
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
71670UVNP UV transilluminator; w/PC, w/o printer; 8-bit, 2.0Mpx camera
16-30D-L Single-wavelength UV, dual-light transilluminator; w/PC and thermal printer; 12-bit, 1.4 Mpx camera
16-30D-LNP Single-wavelength UV, dual-light transilluminator; w/PC, w/o printer; 12-bit, 1.4 Mpx camera
16-30F-DL Single-wavelength UV, dual-light transilluminator; w/PC, thermal printer; 8-bit, 1.4 Mpx cam
16-30F-DLNP Single-wavelength UV, dual-light transilluminator; w/PC; 8-bit, 1.4 Mpx camera
16-30F-UV Single-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 1.4 Mpx camera
16-30F-UVNP Single-wavelength UV transilluminator; w/PC; 8-bit, 1.4 Mpx camera
16-30U-V UV transilluminator; w/PC and thermal printer; 8-bit, 0.4 Mpx camera
16-30U-VNP UV transilluminator; w/PC, w/o printer; 8-bit, 0.4 Mpx camera
71670DLNP Single-wavelength UV, dual-light transilluminator; w/PC, w/o printer; 8-bit, 2.0 Mpx camera
16-70F-DL Single-wavelength UV, dual-light transilluminator; w/PC, thermal printer; 8-bit, 2.0 Mpx
16-70F-DLNP Single-wavelength UV, dual-light transilluminator; w/PC; 8-bit, 2.0 Mpx camera
16-70F-UV Single-wavelength UV transilluminator; w/PC and thermal printer; 8-bit, 2.0 Mpx camera
16-70F-UVNP Single-wavelength UV transilluminator; w/PC; 8-bit, 2.0 Mpx camera
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 71670UVNP Supplier Fotodyne™ Supplier No. 71670UVNP
Includes
8-48mm (6X) manual zoom lens, manual filter cassette, choice of CCD camera, choice of transilluminator, FOTO/FX darkroom with dual white-light and UV epi-illumination, image acquisition software, PCI frame grabber (with 0.4 megapixel camera) or firewire (with 1.4 and 2.0 megapixel cameras) card, Coomassie™ blue filter (dual-light workstations only), PC, optional thermal printer, all necessary cables and connectors
Large-format transilluminators provide the flexibility to capture images of many different sample types, including gels, microplates, fluorescent blots, colorimetric blots, TLC plates, and bacterial plates
  • Systems are designed to be easily upgraded to include a cooled CCD camera for low-light samples
  • Files can be archived in several formats to preserve data integrity for downstream analysis
  • Workstations can be networked to make files more accessible
  • Manual zoom lens allows adjustments to be made by rotating the on-lens aperature, zoom, and focus rings
  • All systems are PC-compatible
FOTO/FX Benchtop Darkroom
  • Light-tight enclosure houses a CCD camera with zoom lens and filters
  • Flat black interior minimizes stray light
  • Gliding transilluminator platform and side-opening door with interlock provide convenient, safe operation of large-format transilluminators
  • Two interior visible epi-light panels allow easy focusing and even illumination of opaque samples
  • Dual UV epi-lights can be used to optimize sensitivity of samples stained with fluorescent dyes
  • Camera mounting system allows for easy upgrading to other cameras
  • Motorized filter wheel permits filters to be selected through computer software or the darkroom touchpad
FOTO/Flex Transillumators
  • Choose single-wavelength UV; single-wavelength UV, dual-light (UV plus white light); dual-wavelength UV; or dual-wavelength UV, dual-light models
  • UV transilluminators have 25 × 28cm UV glass; four 8 watt, 312nm (peak emission) bulbs; UV transparent protective cover; and preparative and analytical intensity settings
  • White-light transilluminators have a proprietary electroluminescent (EL) visible light panel that offers 15 times more uniform light illumination than traditional bulb-style transilluminators and continuously adjustable intensity control
Image Acquisition Software
  • Integration to 8.3 min. in 1/30 sec. increments
  • Live preview
  • Brightness and contrast
  • Image manipulation: invert, zoom, mirror, rotate, annotate (date, time, text, circle, arrow)
  • Image storage in choice of TIF, JPG, BMP, and PNG formats