Learn More
BIOHIPPO FOX01/IRE REPORTER LENT2X10 6
SDP
Supplier:  BIOHIPPO BHV194000363
Encompass
Fluorescent FOX01/IRE Reporter Lentivirus High-quality lentiviral reporter system that provides a sensitive fluorescent readout to FOX01 activity in human/mouse cells. The transcription factor reporter is preferentially activated by FOX01(vs FOX03) and can readout transcription inhibition by FOXP1 and insulin/Akt signaling pathway activation via the IRE. The lentivirus is purified by PEG precipitation and sucrose gradient centrifugation and is ideal for use with difficult-to-transfect cells including primary and/or thawed cells.
Catalog No. NC3165668
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.