Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXA1 Monoclonal Antibody (3A8)
GREENER_CHOICE

Catalog No. PIMA1091 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1091 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1091 Supplier Invitrogen™ Supplier No. MA1091
  • $598.50
    $449.65 / Each of 1

    Save $148.85

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-091 has been successfully used in Western blot and immunohistochemistry (paraffin) applications with human, mouse and canine samples.

This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3A8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene FOXA1
Gene Accession No. P35582, P55317
Gene Alias Ci-fkh; fkd7; fkh; fork head domain; forkhead box A1; forkhead box protein A1; forkhead-7; Foxa1; FoxA4a; hepatocyte nuclear factor 3 alpha (winged helix transcription factor); hepatocyte nuclear factor 3, alpha; hepatocyte nuclear factor 3-alpha; HNF3A; HNF-3A; HNF3alpha; HNF-3-alpha; HNF3-alpha; MGC33105; MGC52590; MGC75967; pintallavis; TCF3A; Tcf-3a; tFoxA1; Transcription factor 3A; xfkh1; xfkh2
Gene Symbols FOXA1
Host Species Mouse
Immunogen Recombinant protein expressed in E.coli corresponding to amino acids 1-473 of human FOXA1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15375, 3169, 490657
Target Species Human, Mouse, Canine
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.