Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXA2 Recombinant Rabbit Monoclonal Antibody (9H5L7)

Catalog No. 701698 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-169-8 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-169-8 Supplier Invitrogen™ Supplier No. 701698
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Horse, Cat and Dog. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

FOXA2 is a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific genes such as albumin and transthyretin. They also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. FOXA2 has been linked to sporadic cases of maturity-onset diabetes of the young. Transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXA2
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 9H5L7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene FOXA2
Gene Accession No. Q9Y261
Gene Alias FKHL20; forkhead box A2; forkhead box protein A2; Foxa2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3 beta (winged helix transcription factor); hepatocyte nuclear factor 3, beta; hepatocyte nuclear factor 3-beta; hepatocyte nuclear factor-3beta; HNF3B; Hnf-3b; HNF3beta; HNF-3BETA; HNF-3-beta; HNF3-beta; MGC19807; putative; RONU; TCF3B; TCF-3B; transcription factor; transcription factor 3B; transcription factor Foxa2; WHN
Gene Symbols FOXA2
Host Species Rabbit
Immunogen Protein corresponding to Human FOXA2 (aa 236-451).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3170
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.