Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA1813 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1813 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA1813 Supplier Invitrogen™ Supplier No. PA1813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-813 detects FOXA3 from human samples. PA1-813 has been successfully used in Western blot and ChIP applications. By Western blot, PA1-813 detects a ~37 kDa band representing FOXA3 from mouse liver. The PA1-813 immunogen is a synthetic peptide corresponding to residues S(4) V K M E A H D L A E W S Y Y P E A G E(23) of mouse FOXA3. This immunizing peptide (PEP-299) is available for use in neutralization and control experiments.

FOXA3 encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. The crystal structure of a similar protein in rat has been resolved.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXA3
Applications ChIP Assay, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene FOXA3
Gene Accession No. P32183, P35584, P55318
Gene Alias FKHH3; Fork head-related protein FKH H3; forkhead box A3; forkhead box protein A3; Foxa3; hepatocyte nuclear factor 3 gamma (winged helix transcription factor); hepatocyte nuclear factor 3, gamma; hepatocyte nuclear factor 3-gamma; HNF3G; HNF-3G; HNF-3-gamma; Tcf3g; Tcf-3g; Transcription factor 3G
Gene Symbols FOXA3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues S(4) V K M E A H D L A E W S Y Y P E A G E(23) of mouse FOXA3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15377, 25100, 3171
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.