Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518170
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518170 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518170 Supplier Invitrogen™ Supplier No. PA518170
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, human, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Foxb1 gene ontology annotations related to this gene include anatomical structure morphogenesis; axon target recognition; cell differentiation; cell migration in diencephalon; epithelial cell differentiation involved in mammary gland alveolus development; floor plate development; hypothalamus cell migration; inferior colliculus development; lactation; mammary gland lobule development; mammillary body development; mammillothalamic axonal tract development; midbrain development; negative regulation of neuron apoptotic process; regulation of transcription by RNA polymerase II; somitogenesis; spinal cord development; telencephalon cell migration; thalamus development; urogenital system development; visual learning.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXB1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Foxb1
Gene Accession No. Q64732, Q99853
Gene Alias C43; FKH5; forkhead box B1; forkhead box B1a; forkhead box B1b; forkhead box protein B1; Foxb1; Foxb1a; Foxb1b; Hfh-e5.1; HFKH-5; mesoderm/mesenchyme forkhead 3; Mf3; Transcription factor FKH-5; Twh
Gene Symbols Foxb1
Host Species Goat
Immunogen Synthetic peptide sequence (TSPASALHSVAVH) corresponding to the C-terminus amino acids of FOXB1 (aa 313-325).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27023, 64290
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.