Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA1807 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1807 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA1807 Supplier Invitrogen™ Supplier No. PA1807
  • $689.59
    $500.65 / Each of 1

    Save $188.94

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-807 detects FOXC1 from human and mouse samples. PA1-807 has been successfully used in Western blot procedures. By Western blot, PA1-807 detects a ~50 kDa band representing FOXC1 from HepG2 cells and mouse kidney cells. This antibody also detects a nonspecific band at ~45 kDa from HepG2 cells, and at ~60 kDa from mouse kidney samples. The PA1-807 immunogen is a synthetic peptide corresponding to residues A(423) V D D P L P D Y S L P(434) of human FOXC1. PA1-807 can be used with blocking peptide PEP-298.

FOXC1 is a protein belonging to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this protein is not known, however, it plays a role in the regulation of FGF19-FGFR4-MAPK pathway to promote both the development and maintenance of anterior segment structures within the eye. Mutations in this gene cause various glaucoma, iridogoniodysgenesis anomaly, Peters anomaly that includes central corneal leukoma, absence of the posterior corneal stroma and Descemetmembrane and Axenfeld-Rieger anomaly characterized by posterior corneal embryotoxon, iris adhesion to the Schwalbe line, hypertelorism, hypoplasia of the malar bones, congenital absence of some teeth and mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXC1
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene FOXC1
Gene Accession No. Q12948, Q61572
Gene Alias ARA; ch; congenital hydrocephalus; Fkh1; fkh-1; Fkhl7; forkhead box C1; forkhead box C1 protein; forkhead box protein C1; forkhead, drosophila, homolog-like 7; forkhead/winged helix-like transcription factor 7; forkhead-related activator 3; forkhead-related protein FKHL7; Forkhead-related transcription factor 3; Foxc1; FREAC3; FREAC-3; frkhda; HGNC:3800; IGDA; IHG1; IRID1; Mesoderm/mesenchyme forkhead 1; Mf1; MF-1; Mf4; myeloid factor-delta; RIEG3; Transcription factor FKH-1
Gene Symbols FOXC1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues A(423) V D D P L P D Y S L P(434) of human FOXC1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17300, 2296
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.