Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524588
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524588 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524588 Supplier Invitrogen™ Supplier No. PA524588
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FOXC2 Polyclonal Antibody for Flow, IHC (P), Western Blot

Forkhead box protein C2 (FOXC2) is a transciptional activator and is characterized by a distinct DNA-binding forkhead domain. FOXC2 may be involved in the formation of special mesenchymal tissues. Mutations affecting the gene can cause lymphedema-distichiasis (LYD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXC2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FOXC2
Gene Accession No. Q99958
Gene Alias BF-3; brain factor 3; Fkh14; Fkhl14; forkhead box C2; forkhead box C2 (MFH-1 mesenchyme forkhead 1); forkhead box C2 (MFH-1, mesenchyme forkhead 1); forkhead box protein C2; forkhead homolog 14; forkhead, Drosophila, homolog-like 14; forkhead-related protein FKHL14; FOXC2; Hfhbf3; HFH-BF-3; LD; mesenchyme fork head protein 1; mesenchyme forkhead 1; MFH1; MFH-1; MFH-1 protein; MFH-1,mesenchyme forkhead 1; Transcription factor FKH-14
Gene Symbols FOXC2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 159-186 amino acids from the central region of human FOXC2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2303
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.