Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXD4L5 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5103845
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103845 Supplier Thermo Scientific Supplier No. PA5103845

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FOXD4L5.

5 FOXD4-like proteins (FOXD4L2, FOXD4L3, FOXD4L4, FOXD4L5 and FOXD4L6) are encoded by a strongly repeated region of the 9q12 chromosome region. They are very similar and it is therefore difficult to provide a clear and unambiguous protein sequence. Our sequences are in agreement with HGNC nomenclature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXD4L5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FOXD4L5
Gene Accession No. Q5VV16
Gene Alias bA15J10.2; forkhead box D4-like 5; forkhead box protein D4-like 5; FOXD4L5; FOXD4-like 5
Gene Symbols FOXD4L5
Host Species Rabbit
Immunogen A synthesized peptide derived from human FOXD4L5(Accession Q5VV16), corresponding to amino acid residues P233-K283.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653427
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.