Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549759 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549759 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549759 Supplier Invitrogen™ Supplier No. PA549759
  • $632.00
    $502.65 / Each of 1

    Save $129.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total FOXE1 protein.

This intronless gene belongs to the forkhead family of transcription factors, which is characterized by a distinct forkhead domain. This gene functions as a thyroid transcription factor which likely plays a crucial role in thyroid morphogenesis. Mutations in this gene are associated with congenital hypothyroidism and cleft palate with thyroid dysgenesis. The map localization of this gene suggests it may also be a candidate gene for squamous cell epithelioma and hereditary sensory neuropathy type I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXE1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FOXE1
Gene Accession No. O00358, Q8R2I0
Gene Alias FKHL15; forkhead box E1; forkhead box E1 (thyroid transcription factor 2); forkhead box E2; forkhead box protein E1; Forkhead box protein E2; forkhead, drosophila, homolog-like 15; forkhead-related protein FKHL15; Foxe1; FOXE2; HFKH4; HFKL5; HNF-3/fork head-like protein 5; NMTC4; Thyroid transcription factor 2; Titf2; TTF2; TTF-2
Gene Symbols FOXE1
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human TTF2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110805, 192274, 2304
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.