Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7204736 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8233 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8233 Supplier Proteintech Group Inc Supplier No. 127641AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FOXG1B belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of FOXG1B has not yet been determined; however, it may play a role in the development of the brain and telencephalon. This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon. This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon.

Specifications

Antigen FOXG1
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FOXG1
Gene Accession No. P55316, Q00939, Q60987
Gene Alias BF 1, BF 2, BF1, BF2, Brain factor 1, Brain factor 2, FHKL3, FKH2, FKHL1, FKHL2, FKHL3, FKHL4, forkhead box G1, Forkhead box protein G1, Forkhead box protein G1A, Forkhead box protein G1B, Forkhead box protein G1C, Forkhead related protein FKHL1, Forkhead related protein FKHL2, Forkhead related protein FKHL3, FOXG1, FOXG1A, FOXG1B, FOXG1C, HBF 1, HBF 2, HBF 3, HBF G2, HBF2, HFK1, HFK2, HFK3, KHL2, QIN
Gene Symbols FOXG1
Host Species Rabbit
Immunogen FOXG1 Fusion Protein Ag3426
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15228, 2290, 24370
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.