Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530031 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530031 Supplier Invitrogen™ Supplier No. PA530031
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, H1299, HCT116, 293T, A431, HeLa, HepG2, FOXI1-transfected 293T, HepG2 nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it is possible that this gene plays an important role in the development of the cochlea and vestibulum, as well as embryogenesis. Mutations in this gene may be associated with the common cavity phenotype. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXI1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene FOXI1
Gene Accession No. Q12951, Q922I5
Gene Alias 5830401E05Rik; Fkh10; FKHL10; Forkhead; forkhead box I1; forkhead box protein I1; forkhead homolog 10; forkhead-like 10; forkhead-related activator 6; forkhead-related protein FKHL10; Forkhead-related transcription factor 6; Foxi1; FREAC6; FREAC-6; Hepatocyte nuclear factor 3 forkhead homolog 3; HFH3; HFH-3; HNF-3/forkhead homolog 3; HNF-3/fork-head homolog 3; HNF-3/fork-head homolog-3; MGC34197
Gene Symbols FOXI1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FOXI1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14233, 2299
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.