Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1802 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1802 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1802 Supplier Invitrogen™ Supplier No. PA1802
  • $648.50
    $500.65 / Each of 1

    Save $147.85

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-802 detects FoxL2 protein in mouse and hamster samples. PA1-802 has successfully been used in Western blot , immunofluorescence and immunohistochemistry procedures. By Western blot, this antibody detects an ~39 kDa protein representing FozL2 in CHO whole cell lysate. The PA1-802 immunogen is a synthetic Peptide corresponding to residues M(1) M A S Y P E P E D T A G T(14) of mouse FoxL2. This sequence is completely conserved in mouse and rat and 92% conserved in human. This peptide (Cat. # PEP-238) is available for use in neutralization and control experiments.

FOXL2 is a winged helix/forkhead transcription factor that is highly conserved in human, goat, mouse, and certain aquatic species. Given its cross-species conservation, it appears to play a critical role evolutionarily, likely in ovarian differentiation in mammals and in ovarian somatic cell differentiation and in further follicle development and/or maintenance. Immunohistochemical data shows that FOXL2 is a nuclear protein specifically expressed in eyelids and in fetal and adult ovarian follicular cells. It does not undergo any major posttranslational modifications. FOXL2 is also found to be associated with blepharo-phimosis/ptosis/epicanthus inverses syndrome (BPES). There are two forms of BPES. Type I BPES shows eyelid abnormal-ities that are associated with ovarian failure. In type II BPES, only eyelid defects are found. Studies have shown that FOXL2 is expressed in the mesenchyme of developing mouse eyelids and in adult ovarian follicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXL2
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene FOXL2
Gene Accession No. O88470, P58012
Gene Alias AU045128; Blepharophimosis; BPES; BPES1; epicanthus inversus; forkhead box L2; forkhead box protein L2; forkhead transcription factor; forkhead transcription factor FOXL2; forkhead transcription factor L2; Foxl2; LUSTR2; MGC14393; Pfrk; P-Frk; PINTO; pituitary forkhead factor; POF3; putative transcription factor foxl2
Gene Symbols FOXL2
Host Species Rabbit
Immunogen Synthetic Peptide corresponding to residues M(1) M A S Y P E P E D T A G T(14) of mouse FoxL2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26927, 668
Target Species Human, Mouse, Hamster
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.