Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO1/3/4-pan (phospho Thr24/32) Antibody, ProSci
SDP

Catalog No. 16818776
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-818-776 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-818-776 Supplier ProSci LLC Supplier No. 797880100
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FOXO1/3/4-pan (phospho Thr24/32) Polyclonal Antibody specifically detects Bax (phospho Thr167) in Human, Mouse, Rat Samples. It is validated for Western Blot.

Specifications

Antigen FOXO1/3/4-pan (phospho Thr24/32)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot: 1:500~1:1000
Formulation Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Gene Accession No. NP_002006.2
Gene Alias FKHR, FOXO1, FKHR2, AFX, MLLT7
Host Species Rabbit
Immunogen FOXO1/3/4-pan (phospho Thr24/32) antibody was raised against a peptide sequence around phosphorylation site of threonine 24/32 (S-C-T (p)-W-P) derived from Human FOXO1/3/4-pan .
Purification Method Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2308;4303
Test Specificity The antibody detects endogenous levels of FOXO1/3/4-pan only when phosphorylated at threonine 24/32.
Target Species Human, Mouse, Rat
Content And Storage Store antibody at -20°C for up to one year.
Form Liquid
Isotype IgG
Research Category Phospho-Specific
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.