Learn More
Novus Biologicals™ FoxP3

Description
- RetroMax system takes advantages of two properties of 293 cells (high level of transfectibility and strong E1A-mediated stimulation of CMV promoter controlled transcription)
- 293 cells of nonmurine origin avoid problem of selective packaging and transfer of VL30 genomes (present in all murine packaging cells)
- Vector supernatants are free of helper virus and are of sufficiently high titer within 2 days of transient transfection in 293 cells
- For amplification, transform DH-5alpha or XL-1 Blue and plate on LB plates containing 50mg/mL ampicillin
- Buffer: 20μL 1x TE (10mM Tris, pH 7.5, 1mM EDTA)
Recommended Storage
Store at -20°C. Avoid freeze-thaw cycles.
Specifications
Specifications
| Storage Buffer | 20µL 1x TE (10mM Tris, pH 7.5, 1mM EDTA) |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 2 μg |
| Vector | FoxP3 Vector |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.