Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP3 Antibody (3G3) [Alexa Fluor Plus 555], R&D Systems™
SDP

Catalog No. RD10009442
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10009442 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10009442 Supplier R&D Systems Supplier No. NB10056582AFP555

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems™. Mouse Monoclonal Antibody

FoxP3 Monoclonal antibody specifically detects FoxP3 in Human, Mouse samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen FoxP3
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3G3
Conjugate Alexa Fluor Plus 555
Formulation 50mM Sodium Borate
Gene Alias AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked
Host Species Mouse
Immunogen Full-length His-tagged murine FOXP3 was used as the immunogen. The precise epitope is not known, but it has been mapped to the N-terminal portion of the protein (NP_001107849).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Regulatory Immunology, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 50943
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.