Learn More
Invitrogen™ FOXP3 Monoclonal Antibody (FJK-16s), Brilliant Violet™ 605, eBioscience™, Invitrogen™
Description
Specifications
Specifications
| Antigen | FOXP3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | FJK-16s |
| Concentration | 0.2 mg/mL |
| Conjugate | Brilliant Violet 605 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | Foxp3 |
| Gene Accession No. | Q99JB6 |
| Gene Alias | AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.