Learn More
FOXP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
This antibody is a rabbit polyclonal antibody raised against a peptide mapping within human FOXP3.
Specifications
Specifications
| Antigen | FOXP3 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.29 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FOXP3 |
| Gene Accession No. | Q99JB6, Q9BZS1 |
| Gene Alias | AIID, DIETER, forkhead box P3, Forkhead box protein P3, FOXP3, IPEX, JM2, PIDX, Scurfin, XPID |
| Gene Symbols | Foxp3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.