Learn More
Invitrogen™ eBioscience™ Foxp3 / Transcription Factor Staining Buffer Set
Catalog No. 501128857
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
50-112-8857 1 kit
1 options
Contains Sodium azide and formaldehyde
Catalog No. 50-112-8857 Supplier Invitrogen™ Supplier No. 00552300
Only null left
Add to Cart
Add to Cart
Contains Sodium azide and formaldehyde
Formulated and optimized for staining with antibodies against transcription factors and nuclear proteins (e.g. Foxp3 and Ki-67), cytokines and chemokines
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kit
Reagent Type Fixation Diluent, Fixation Reagent, Permeabilization Diluent, Permeabilization Reagent
Product Line eBioscience
Product Type Staining Buffer
For Research Use Only. Not for use in diagnostic procedures.