Learn More
Invitrogen™ eBioscience™ Foxp3 / Transcription Factor Fixation/Permeabilization Concentrate and Diluent
Catalog No. 501129060
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
50-112-9060 1 kit
1 options
Contains Sodium azide and formaldehyde
Catalog No. 50-112-9060 Supplier Invitrogen™ Supplier No. 00552100
Add to Cart
Edge
Add to Cart
Contains Sodium azide and formaldehyde
For staining with antibodies against transcription factors and nuclear proteins (e.g. Foxp3 and Ki-67), cytokines and chemokines. Should be used with the Permeabilization Buffer. All three components sold together as the Foxp3 / Transcription Factor Staining Buffer Set.
The Foxp3 Transcription Factor Fixation/Permeabilization Concentrate and Diluent solutions for staining with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67, as well as cytokines and chemokines. It should be used in combination with the Permeabilization Buffer (10X) (cat. 00-8333). All three components can be purchased together in the Foxp3 / Transcription Factor Staining Buffer Set (cat. 00-5523).
Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kit
Reagent Type Fixation Diluent, Fixation Reagent, Permeabilization Diluent, Permeabilization Reagent
Product Line eBioscience
Product Type Fixation/Permeabilization Diluent
For Research Use Only. Not for use in diagnostic procedures.