Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXQ1 Antibody (OTI4D9), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288222
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288222 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288222 Supplier R&D Systems Supplier No. NBP272338APC

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

FOXQ1 Monoclonal specifically detects FOXQ1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen FOXQ1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4D9
Conjugate APC
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation PBS
Gene Alias forkhead box Q1, Hepatocyte nuclear factor 3 forkhead homolog 1, HFH-1, HFH1forkhead box protein Q1, HNF-3/forkhead-like protein 1, winged helix/forkhead transcription factor
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 11-295 of human FOXQ1(NP_150285) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 94234
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.