Learn More
Medchemexpress LLC HY-P1416A 1mg Medchemexpress, Foxy-5 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1416A1MG
Encompass_Preferred
Medchemexpress, HY-P1416A 1mg Foxy-5 (TFA) CAS: Foxy-5 is a WNT5A mimicking peptide with anti-metastatic effect, which lacks a heparan sulfate-binding domain. Foxy-5 triggers cytosolic free calcium signaling without affecting β-catenin activation and it impairs the migration and invasion of epithelial cancer cells [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4196
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.