Learn More
MSA™ FP Diamond™ Energy-Absorbing Lanyard
Catalog No. 191600199 Shop All MSA Products
Change view
Click to view available options
Connector Types:
HL2000
Two GL3100
Two HL2000
Two HL64
Leg Configuration:
Double-Leg
Single-Leg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types Leg Configuration
19-160-0199 Two GL3100 Double-Leg
19-160-0197 Two HL64 Double-Leg
19-160-0198 Two HL64 Single-Leg
19-130-3894 HL2000 Single-Leg
19-160-0196 Two HL2000 Double-Leg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-160-0199 Supplier MSA™ Supplier No. 10047290
Lightweight and streamlined design
Connects the back (dorsal) D-ring of MSA full-body harnesses to a qualified anchorage and anchorage connector
  • Keeps average fall arrest forces below 900 lb.
  • Durable polyester sheath for increased abrasion and UV resistance
  • Pouchless design for lighter weight and more comfortable wear
  • Aluminum or zinc-plated steel snaphooks with auto-close and lock
  • Highly visible deployment tag
  • Swivel HL200 harness connection
  • Single-leg configuration is bi-directional to make training easier, while twin-leg configuration allows for 100% tie off
Compliance
US: ANSI/ASSE Z359.13, Z359.3; OSHA 29 CFR 1910.66, ASTM F887-005; CAN: Z259.11-05
Specifications
Material Polyester
Leg Configuration Double-Leg
Length (English) 6 ft.
Connector Types Two GL3100
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.