Learn More
Agilent Technologies FP SEAHRSE 384 140U PE 25 10CS
Supplier:  Agilent Technologies 201035100
Encompass
Seahorse polypropylene 384-well filter microplate with 25um high molecular weight polyethylene membrane, 140ul/well, long drip, case of 10, recommended receiver plate 201274-100
Catalog No. 50-995-958
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.