Learn More
TARGETMOL CHEMICALS INC FPL64176 10MG
Supplier: TARGETMOL CHEMICALS INC T2278810MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FPL64176 is a L-type calcium channels activator. purity: 97%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.