Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FPR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675527 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-527 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-527 Supplier Invitrogen™ Supplier No. PA141398
  • $585.00
    $478.65 / Each of 1

    Save $106.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Suggested positive control: human liver protein, liver lysate. Immunogen sequence: CTFNFSPWTN DPKER.

FPR1 is a member of chemoattractant/chemokine GPCRs expressed mainly on leukocytes, neutrophils and monocytes. FPR1 is a seven transmembrane pertussis toxin-sensitive GPCR and binds to formylated peptides generated by bacteria resulting in leukocyte trafficking to the site of inflammation. Ligand binding to FPR results in activation of multiple effectors including phospholipase C leading to calcium mobilization and PKC activation. Other implications of FPR signaling include polarization of actin cytoskeleton, activation of various integrins, increased cell migration, phagocytosis, degranulation and release of reactive oxygen intermediates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FPR1
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene FPR1
Gene Accession No. P21462, P33766
Gene Alias fMet-Leu-Phe receptor; fMLF-R; FMLP; fMLP receptor; formyl peptide receptor 1; FPR; Fpr receptor; FPR1; FPR-1; lipoxin A4 receptor; LXA4R; N-formyl peptide receptor; N-formylpeptide chemoattractant receptor
Gene Symbols FPR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 176-190 of human FPR1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14293, 2357
Target Species Human, Mouse, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.