Learn More
TARGETMOL CHEMICALS INC FR-167356 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T27371
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. FR-167356 is a specific inhibitor of a3 isoform vacuolar type H⁺-ATPase with IC50s of 170 nM 370 nM and 220 nM for osteoclast plasma membranes renal brush border membranes and macrophage microsomes. FR-167356 reduces bone metastasis of B16-F10 cells. purity: 99%
Catalog No. 50-258-0622
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.