Learn More
National Safety Apparel™ FR Classic Cotton™ Long Sleeve T-Shirt
Shop All National Public Seating Corporation Products
Description
- CAT 2
- Fabric/Material: 6.25 oz. FR Classic Cotton™
- Soft, breathable fabric
- Reinforced left chest pocket
- Generous sizing for optimal movement
- Increased comfort at neckline with tagless label printing and covered seams
- Unique color options
- Home laundry
- Quickly identify compliance with exterior labeling
- FR fabric will not melt, drip, or ignite
- Made in the USA
Specifications
Specifications
| Size | Medium |
| Material | Cotton |
| Color | Navy |
| Sleeve Style | Long Sleeve |
| Arc Flash Rating | 12 cal/cm2 |
| Certifications/Compliance | NFPA 70E/ASTM F1506/CSA Z462 for Arc Flash, OSHA 1910.269 |