Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ FR Control™ Long Underwear
SDP

Catalog No. 17355111 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Color:
Desert Sand
Navy
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
17-355-111 Desert Sand Small
17-355-105 Navy Small
17-355-106 Navy Medium
17-355-107 Navy Large
17-355-108 Navy X-Large
17-355-109 Navy 2X-Large
17-355-110 Navy 3X-Large
17-355-112 Desert Sand Medium
17-355-113 Desert Sand Large
17-355-114 Desert Sand X-Large
17-355-115 Desert Sand 2X-Large
17-355-116 Desert Sand 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 17-355-111 Supplier National Safety Apparel™ Supplier No. U52JKSRSM
Add to Cart
Add to Cart
This item is not returnable. View return policy

Long underwear constructed of lightweight, breathable, moisture-wicking fabric features heat stress reduction.

  • Proactively regulates skin temperature (± 3 degrees)
  • Reduces perspiration
  • Maintains a comfortable work environment
  • Keeps the wearer cooler on hot days, warmer on cool days
  • Heat stress reduction
  • Odor control technology
  • Lightweight, breathable fabric
  • Unsurpassed moisture wicking and drying rate
  • FR fabric will not melt, drip, or ignite
  • Inherently flame resistant
  • FR protection will not wash/wear out
  • 4-way stretch fabric for mobility and recovery
  • Made in the USA

Specifications

Size Small
Gender Unisex
Product Type Multirisk Pants
Material Modacrylic
Color Desert Sand
Seam Type Stitched
No. of Pockets None
Closure Type Pull On
Disposable Reusable
Hazard Risk Category CAT 1
Arc Flash Rating 4 cal/cm²
Certifications/Compliance NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Style Base Layer Pant
For Use With (Application) Utilities, Electrical Contractors, Manufacturing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.