Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's FR/CP Lab Coat

Catalog No. 19800061 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
Small
Medium
X-Large
2X-Large
3X-Large
Style:
Regular
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Style
19-800-061 3X-Large Regular
19-800-055 Small Regular
19-800-053 Medium Regular
19-800-057 X-Large Regular
19-800-059 2X-Large Regular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-800-061 Supplier Bulwark™ FR Supplier No. KNR2RB 3L 0R

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides dual hazard protection with flame resistance and chemical protection. Bulwark™ FR FR/CP Lab Coats are lightweight, breathable, and comfortable to wear. They employ Nomex™ IIIA fabric to prevent ignition and ShieldCXP™ to repel small amounts of liquid chemicals.

Provides dual hazard protection with flame resistance and chemical protection. Bulwark™ FR Lab Coats are lightweight, breathable, and comfortable to wear. They employ Nomex™ IIIA fabric to prevent ignition and ShieldCXP™ to repel small amounts of liquid chemicals.

  • Nomex™ IIIA fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame
  • ShieldCXP repels small amounts of liquid chemicals that are either dropped or splashed on the fabric
  • Nomex™ knit cuffs are fire resistant but do not provide chemical protection - these are typically covered by gloves
  • Black lay-down collar
  • Concealed snap front closure
  • Splash-resistant side pass-through for easy inner pocket access
  • Pockets include one double-stitched chest pocket and two large double-stitched lower front pockets with snap flaps
  • Available in a variety of sizes with regular and long variants

Certifications

NFPA 2112 UL-Certified

Compliance

Meets NFPA 2112 Standard on Flame Resistance Clothing for Protection of Industrial Personnel Against Short-Duration Thermal Exposure from Fire. UL certified.

Specifications

Size 3X-Large
Gender Men's
Material Nomex IIIA
Color Royal Blue
Closure Type Snaps
Cuff Style Knit
Style Regular
No. of Pockets 3
Finish Westex ShieldCXP Chemical Splash Protection
Size (English) Chest 56.5 in.
Size (Metric) Chest 143.5 cm
Length (English) Sleeve 39.75 in.
Length (Metric) Sleeve 100.3 cm
Weight (English) 4.5 oz.
Weight (Metric) 128 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.