Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ FR Food Processing Lab Coat
SDP

Catalog No. 17355102 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-355-102 X-Large
17-355-099 Small
17-355-100 Medium
17-355-101 Large
17-355-103 2X-Large
17-355-104 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-355-102 Supplier National Safety Apparel™ Supplier No. C09UJLCFSXL42
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lightweight and breathable food processing lab coat provides arc flash and flash fire protection.

  • Created for food service: no buttons, no loose or hanging components
  • Side pass-through pockets
  • Generous cut allows for unrestricted movement
  • Covered snaps on front and cuffs
  • Dual certified for arc flash and flash fire protection
  • Lightweight and breathable design
  • Standard coat length is 42 in.
  • Made in the USA

Specifications

Product Type Flame-resistant Lab Coat
Size X-Large
Gender Unisex
Material FR Cotton
Color Navy
Closure Type Snaps
Cuff Style Hemmed
Disposable Reusable
Sterility Non-sterile
Resistance Flame
Style Standard Length
No. of Pockets 2
Model C09UJLCFS
Hazard Rating CAT 2
Arc Flash Rating 8 cal/cm²
Certifications/Compliance NFPA 70E, NFPA 2112, ASTM F1506, CSA Z462, OSHA 1910.269
For Use With (Application) Food Processing, Manufacturing
Length (English) 42 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.