Learn More
Bulwark™ FR Lab Coats, Knit Cuffs
Catalog No. 17004364 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Style:
Regular
Tall
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Style
17-004-364 X-Small Regular
17-004-365 Small Regular
17-004-366 Medium Regular
17-004-367 Large Regular
17-004-368 X-Large Regular
17-004-369 2X-Large Regular
17-004-370 3X-Large Regular
17-004-371 4X-Large Regular
17-004-372 5X-Large Regular
17-004-373 Medium Tall
17-004-374 Large Tall
17-004-375 X-Large Tall
17-004-376 2X-Large Tall
17-004-377 3X-Large Tall
17-004-378 4X-Large Tall
17-004-379 5X-Large Tall
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 17-004-364 Supplier Bulwark™ Supplier No. KNC2RB XS 0R
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lab coat designed for functionality, comfort, and safety with a CAT 1 arc rating and NFPA 2112 compliance.
  • Certifications Level 1: NFPA 2112, ASTM F1506
  • Certifications Level 2: Category 1 Protection
  • Hazard: Arc Flash, Flash Fire
  • Wash Care: Industrial Laundry - Light Soil
  • Rib cuffs for protection during movement
  • Concealed snap front closure to avoid scratching surfaces
  • Four front mitered style pockets for ample storage
  • Pass-through at hips to access underneath
  • Protection: ATPV Arc Rating: 4.4
  • Country of Origin: Imported, Made in USA Fabric
Specifications
Product Type Flame-resistant Lab Coat
Size X-Small
Gender Men's
Material Nomex
Color Royal Blue
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Flame
Style Regular
No. of Pockets 4
Model KNC2RB
Arc Flash Rating 4.4 Calories/cm2
Certifications/Compliance NFPA 2112, ASTM 1506
Construction Sewn