Learn More
Cytiva Frac30 Model Fraction Collector
Catalog No. 29023051 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Akta Start Purification System
Catalog No. For Use With (Equipment)
29-023-051 Akta Start Purification System
1 options
Catalog No. 29-023-051 Supplier Cytiva Supplier No. 29023051
This product is not available for purchase online. For additional information, please contact your Fisher Scientific Technical Sales Representative.
Used as a round fraction collector. Cytiva Frac30 Model Fraction Collector allows you to collect up to 30 fractions.
  • Controlled via the AKTA protein purification system start touch screen instrument display
  • Also controlled by using UNICORN start software via computer
  • Collect up to 30 fractions
  • Supports four tube sizes, ranging from 1.5mL to 15mL
  • Fractions can be collected in volumes ranging from 0.5mL to 15mL
  • The entire fraction collector rack can be transferred as required
TRUSTED_SUSTAINABILITY
Specifications
Type Fraction Collector
For Use With (Equipment) Akta Start Purification System