Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fraction Pie Puzzle
SCI_ED

Catalog No. S96088 Shop All Educational Insights Products
Change view
Click to view available options
Class Size:
1 to 2 players
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96088 1 to 2 players
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CHOKING HAZARD —Small parts. Not for children under 3 yrs.
Catalog No. S96088 Supplier Educational Insights Supplier No. EI8445
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHOKING HAZARD —Small parts. Not for children under 3 yrs.

Includes

  • Game board, leveled puzzle cards (50), foam fraction pieces (65), guide, and solutions for every puzzle

A puzzle and a conceptual fraction lesson, all rolled into fun

  • An entertaining way for students to understand fractions
  • Reinforces basic fraction concepts and teaches students how fractional parts make up a whole
  • Demonstrates quivalencies, common denominators, and multiples
  • Select a card, place the starting pieces, and then complete the circles with the remaining fraction pie pieces
  • Leveled from easy to difficult and self-correcting, the puzzles are perfect as a small group or center activity
  • Class size: 1-2 players

Order Info

For one to two players

Specifications

For Use With (Application) Reinforces basic fraction concepts
Includes Game board, leveled puzzle cards (50), foam fraction pieces (65), guide, and solutions for every puzzle
Class Size 1 to 2 players
Description Fraction Pie Puzzle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.