Learn More
United Scientific Supplies Fractional Brass Masses
SDPSCI_ED
Catalog No. S40200 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
12 Piece Set
Catalog No. Quantity
S40200 12 Piece Set
1 options
Catalog No. S40200 Supplier United Scientific Supplies Supplier No. FWB500
Add to Cart
Add to Cart
Mass set meets weighing standards.
Set comes in compact storage case and includes masses from 500mg to 1g.
  • Individual masses for calibration
  • Build your own customized set
  • Replace missing or damaged masses
Specifications
Product Type 12-Piece Set, With Lid
Material Nickel Plated Brass
Weight (Metric) 500 mg, 100 mg, 50 mg, 10 mg, 5 mg, 1 mg; Two Each: 200 mg, 20 mg, 2 mg
Includes 12-Piece Set, Compact storage case
Quantity 12 Piece Set