Learn More
Bio Rad Laboratories FRAME-SEAL SLIDE CHAMBERS
Supplier:  Bio Rad Laboratories SLF0601
Encompass
Chamber, Slide; Bio-Rad; Frame-Seal; For incubation; Used with plain, unprinted glass slides; 15 x 15mm; Capacity: 65uL; Include coverslips; 100/pk.
Catalog No. 50-901-3177