Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Franck-Hertz Apparatus II
SDPSCI_ED

Catalog No. S43701 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
12 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S43701 12 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43701 Supplier United Scientific Supplies Supplier No. FHA002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Franck-Hertz experiment yields basic evidence for the existence of quantum states.

  • The Franck-Hertz Apparatus II is an improved model of the well-known Franck-Hertz Apparatus (FHA001), featuring stable, updated electronics and robust digital readouts for voltage and current.
  • New fully shielded coax cables reduce electrical noise.
  • The Franck-Hertz Experiment yields basic evidence for the existence of quantum states.
  • Inside a vacuum tube filled with a trace of Ar gas, a beam of electrons is generated and accelerated towards a counter electrode.
  • A voltage barrier ahead of the counter electrode only allows electrons with a certain minimum energy to get through and make a detectable current.
  • As the accelerating energy is increased, changes in the current are observed.
  • On the way to the counter electrode, electrons collide with Ar atoms.
  • As the electron energy rises, it gives one quantum of energy to the Ar for ionization.
  • Now the electron can't pass the voltage barrier and the current drops.
  • Valleys in the current curve are evidence of the donations of a quantum of energy.
  • Ar-filled Franck-Hertz tube achieves 5-7 current maxima.
  • Anode current range 0.1 nA—1 μA.
  • Automatic scan to oscilloscope or chart recorder, or manual scan.
  • Adjustable, stabilized filament, grid, and anode voltages.
  • 3-1/2 digit voltage and current readouts.
  • Requires user-supplied dual-channel oscilloscope for automatic scan.

Specifications

Grade 12 to 15
Format Kit
Product Type Franck-Hertz Apparatus II
Languages English
Subject Advanced Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.