Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frataxin Mouse anti-Human, Unconjugated, Clone: 4F9, MilliporeSigma™

Catalog No. MABN23132MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN23132MI 25 μg
MABN23131MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN23132MI Supplier MilliporeSigma Supplier No. MABN231325UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Frataxin Monoclonal antibody specifically detects Frataxin in Human samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Frataxin
Applications Western Blot
Classification Monoclonal
Clone 4F9
Conjugate Unconjugated
Gene Accession No. NP_000135
Gene Alias EC:1.16.3.1;Friedreich ataxia protein;Fxn
Gene Symbols FXN, FRDA, X25
Host Species Mouse
Immunogen Full-length human recombinant Frataxin.
Molecular Weight of Antigen ∼14 and 18 kDa observed; 23.13 kDa calculated (full length). Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 4F9 is a mouse monoclonal antibody that specifically detects mature and intermediate forms of human Frataxin.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.